Learn More
StatLab™ Formical-2000 Decalcifier
A fast yet gentle bone decalcifier.
$50.82 - $505.25
Specifications
| Solution Type | Decalcifier |
|---|
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
STL131432
|
StatLab™
131432 |
1 Quart |
Each for $50.82
Case of 4 Each for $203.28
|
|
|||||
|
STL13141
|
StatLab™
13141 |
1 Gallon |
Each for $93.79
Case of 4 Each for $375.16
|
|
|||||
|
STL13145
|
StatLab™
13145 |
5 gal. Cube |
Each for $505.25
|
|
|||||
Description
- Formulated from semiconductor grade formic acid, highly purified reagent grade water and EDTA
- Works for all routine histological specimens, as well as IHC
- Will safely decalcify bone without destroying antigen sites
Specifications
| Decalcifier |