missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid : Triethylamine 1:1 Solution, Honeywell™ Fluka™

for HPLC, 2M:2M concentrate

$1103.92 - $1103.92

Chemical Identifiers

CAS 585-29-5
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00134943
InChI Key PTMFUWGXPRYYMC-UHFFFAOYSA-N
Synonym unii-un4t038p8m, triethylamine formate, teaf, triethylammonium formate, triethylamine, formate, triethylammonium formate mi, triethylammoniumformate, triethylammonium format, hcooh et3n, formic acid triethylamine
PubChem CID 11084032
IUPAC Name N,N-diethylethanamine;formic acid
SMILES CCN(CC)CC.C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044616
View Documents
Honeywell-Fluka
09752100ML
100 mL Glass Bottle
Each for $1,103.92
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

585-29-5
147.218
PTMFUWGXPRYYMC-UHFFFAOYSA-N
11084032
CCN(CC)CC.C(=O)O
C7H17NO2
MFCD00134943
unii-un4t038p8m, triethylamine formate, teaf, triethylammonium formate, triethylamine, formate, triethylammonium formate mi, triethylammoniumformate, triethylammonium format, hcooh et3n, formic acid triethylamine
N,N-diethylethanamine;formic acid

Specifications

585-29-5
C7H17NO2
MFCD00134943
4150472
PTMFUWGXPRYYMC-UHFFFAOYSA-N
2M:2M concentrate
147.218
147.22g/mol
1.01g/mL at 20°C
Glass Bottle
Formic Acid: Triethylamine 1:1 Solution
Colorless
(CH3CH2)3NH+ HCOO-
UN1760
unii-un4t038p8m, triethylamine formate, teaf, triethylammonium formate, triethylamine, formate, triethylammonium formate mi, triethylammoniumformate, triethylammonium format, hcooh et3n, formic acid triethylamine
CCN(CC)CC.C(=O)O
N,N-diethylethanamine;formic acid
11084032
HPLC
100 mL
Liquid
Videos
Safety and Handling

Safety and Handling

P261-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233

H302 + H312-H314-H331-H335

DOTInformation : 8/PGIII

SDS
Documents

Documents