missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid : Triethylamine 1:1 Solution, Honeywell™ Fluka™

Catalog No. 60044616
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044616 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60044616 Supplier Honeywell Chemicals Supplier No. 09752100ML
Only null left

for HPLC, 2M:2M concentrate

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 585-29-5
Molecular Formula C7H17NO2
Molecular Weight (g/mol) 147.218
MDL Number MFCD00134943
InChI Key PTMFUWGXPRYYMC-UHFFFAOYSA-N
Synonym unii-un4t038p8m, triethylamine formate, teaf, triethylammonium formate, triethylamine, formate, triethylammonium formate mi, triethylammoniumformate, triethylammonium format, hcooh et3n, formic acid triethylamine
PubChem CID 11084032
IUPAC Name N,N-diethylethanamine;formic acid
SMILES CCN(CC)CC.C(=O)O

Specifications

Color Colorless
Linear Formula (CH3CH2)3NH+ HCOO-
UN Number UN1760
Beilstein 4150472
Concentration 2M:2M concentrate
Formula Weight 147.22g/mol
Grade HPLC
Density 1.01g/mL at 20°C
Quantity 100 mL
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Formic Acid: Triethylamine 1:1 Solution
Show More Show Less
Hazard Statement P261-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
Precautionary Statement H302 + H312-H314-H331-H335
DOTInformation 8/PGIII