missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid ACS, 98%, MilliporeSigma™
Supplier: Technidata FX044021
Specifications
| Formic Acid | |
| 98% | |
| HCOOH | |
| BDAGIHXWWSANSR-UHFFFAOYSA-N | |
| formic acid | |
| 284 | |
| 46.3 | |
| 25 L | |
| Metal Drum w/Bag |
| 64-18-6 | |
| CH2O2 | |
| methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
| C(=O)O | |
| 46.025 | |
| CHEBI:30751 | |
| ACS/NF/USP | |
| 50°C(122°F) | |
| Liquid |
Chemical Identifiers
| 64-18-6 | |
| 46.025 | |
| methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
| CHEBI:30751 | |
| C(=O)O |
| CH2O2 | |
| BDAGIHXWWSANSR-UHFFFAOYSA-N | |
| 284 | |
| formic acid |