missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid ACS, 98%, MilliporeSigma™
Specifications
Specifications
| Assay Percent Range | 98% |
| Linear Formula | HCOOH |
| Formula Weight | 46.3 |
| Grade | ACS/NF/USP |
| Quantity | 1 L |
| Flash Point | 50°C(122°F) |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Chemical Name or Material | Formic Acid |