missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, ACS, 88%, Spectrum™ Chemical

Catalog No. 18610666
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610666 500 mL Amber Glass Bottle
18610663 100 mL Amber Glass Bottle
18610664 2.5 L Glass Bottle
18610661 4 L Poly-Coated Glass Bottle
18610662 20 L Poly Drum
18610665 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610666 Supplier Spectrum Chemical Mfg Cor Supplier No. F1090500MLGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-18-6 , 7732-18-5

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Amber Glass Bottle
Residue after Evaporation 2e-05
Percent Purity 88%
Grade ACS

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.