missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, ACS, 88%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Percent Purity | 88% |
| Grade | ACS |
| Quantity | 100 mL |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 2e-05 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.