missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 90%, Viscosity Grade, Reagents

Catalog No. 01337595
Change view
Click to view available options
Quantity:
4 L
20 L
15 gal.
200 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337595 15 gal.
01337594 200 L
01337593 20 L
01337592 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337595 Supplier Labchem Inc Supplier No. C210505015E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formic Acid, 90%, Viscosity Grade, Viscosity, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name formic acid
SMILES OC=O

Specifications

pH 2.2
Melting Point 8°C
Boiling Point 106°C
Color Colorless
Quantity 15 gal.
Packaging Poly Drum
Physical Form Liquid
Specific Gravity 1.22
Chemical Name or Material Formic Acid, 90%, Viscosity Grade