missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 90%, Purified, Reagents

Catalog No. p-200020995
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337599 100 mL
01337598 2.5 L
01337597 4 L
01337596 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337599 Supplier Labchem Inc Supplier No. C2105000100A
Only null left

Formic Acid, 90%, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name formic acid
SMILES OC=O

Specifications

pH <2
Melting Point 8°C
Boiling Point 106°C
Color Colorless
Grade Reagent
Quantity 100 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.22
Chemical Name or Material Formic Acid, 90%, Purified