missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 88%, ACS Reagent, Reagents

Catalog No. 01338177
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338177 2.5 L
01338176 4 L
01338175 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338177 Supplier Labchem Inc Supplier No. C11050002.5D1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formic Acid, 88%, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name formic acid
SMILES OC=O

Specifications

pH <2
Melting Point 8°C
Boiling Point 106°C
Color Colorless
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.22
Grade ACS
Chemical Name or Material Formic Acid, 88%, ACS Reagent