missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 60% w/v, Reagents

Catalog No. 01335190
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335190 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335190 Supplier Labchem Inc Supplier No. CF1463004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formic Acid, 60% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name formic acid
SMILES OC=O

Specifications

pH <2
Boiling Point 100°C
Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.1237
Percent Purity 60%
Grade Reagent
Chemical Name or Material Formic Acid, 60% w/v