missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 40% (w/w), Ricca Chemical

Catalog No. R321800010F
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R321800010F 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R321800010F Supplier Ricca Chemical Company Supplier No. R321800010F

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) Mixture
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

CAS Min % 59.0, 39.0
Color Colorless
CAS Max % 61.0, 41.0
Percent Purity 40% w/w
Grade Laboratory
Concentration or Composition (by Analyte or Components) 40% (w/w) Aqueous Solution
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Chemical Name or Material Formic Acid
ShelfLife 24 months