missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid, 2% (w/v), Ricca Chemical

Catalog No. 32135
Change view
Click to view available options
Quantity:
20 L
Packaging:
Cubitainer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
32135 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32135 Supplier Ricca Chemical Company Supplier No. 32135

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) Mixture
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES OC=O

Specifications

CAS Min % 2.16, 96.0
Color Colorless
CAS Max % 2.25, 100.0
Concentration 2% (w/v)
Percent Purity 2% w/v
Grade Laboratory
Quantity 20 L
Packaging Cubitainer
Physical Form Liquid
Chemical Name or Material Formic Acid