Learn More
Mopec™ Formaway Waste Collection Container
Formaway Collection Containers are used to collect spent formalin for neutralizing.
$73.82 - $83.48
Specifications
| Color | White |
|---|---|
| Autoclavable | Not Autoclavable |
| Material | Plastic |
| Product Type | Waste Container |
Description
- Collect waste in a designated collection container
- Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste
- Add the entire contents of one Formaway bottle to each gallon of aldehyde waste
- Secure lid on the collection container and agitate container to thoroughly mix solutions, make sure the agitation ultimately results in complete dissolution of Formaway Formalin Neutralizer Powder
- Allow the mixed solution to stand for 20 to 25 minutes for the reaction to be complete
- If required, test treated solution for any aldehyde residual prior to disposal (follow instructions for Formaway Formaldehyde Residual Screening Test Kit)
- Pour treated waste into sanitary sewer and rinse container with cold tap water, preparing it for reuse
Specifications
| White | |
| Not Autoclavable | |
| Plastic | |
| Waste Container |