Learn More
Mopec™ Formaway Waste Collection Container
Description
- Collect waste in a designated collection container
- Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste
- Add the entire contents of one Formaway bottle to each gallon of aldehyde waste
- Secure lid on the collection container and agitate container to thoroughly mix solutions, make sure the agitation ultimately results in complete dissolution of Formaway Formalin Neutralizer Powder
- Allow the mixed solution to stand for 20 to 25 minutes for the reaction to be complete
- If required, test treated solution for any aldehyde residual prior to disposal (follow instructions for Formaway Formaldehyde Residual Screening Test Kit)
- Pour treated waste into sanitary sewer and rinse container with cold tap water, preparing it for reuse
Specifications
Specifications
| Capacity (English) | 2.5 gal. |
| Color | White |
| Autoclavable | Not Autoclavable |
| Material | Plastic |
| Product Type | Waste Container |