missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formate Standard For Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC ASHCO292X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| HCOO- | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Formate Standard | |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| Sodium formate,distilled water,conductivity or of similar purity | |
| Ion Chromatography | |
| Standard Solution | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| C(=O)[O-].[Na+] | |
| sodium;formate | |
| 500 mL | |
| CHEBI:62965 |
| For Ion Chromatography | |
| 141-53-7,7732-18-5 | |
| 0.15,99.85 | |
| CHNaO2 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Certified Reference Material | |
| Sodium Formate | |
| Water | |
| 68.007 | |
| 2723810 | |
| 1000mg/L |
Chemical Identifiers
| 141-53-7 | |
| 68.007 | |
| 2723810 | |
| sodium;formate |
| CHNaO2 | |
| HLBBKKJFGFRGMU-UHFFFAOYSA-M | |
| CHEBI:62965 | |
| C(=O)[O-].[Na+] |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only