missing translation for 'onlineSavingsMsg'
Learn More

Formamidine Disulfide Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7129460
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00465G 5 g
F004625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00465G Supplier TCI America Supplier No. F00465G
Only null left

Chemical Identifiers

CAS 14807-75-1
Molecular Formula C2H8Cl2N4S2
Molecular Weight (g/mol) 223.13
MDL Number MFCD00012586
InChI Key BFJQSCVWXZOXGK-UHFFFAOYSA-N
Synonym formamidine disulfide dihydrochloride, formamdine disulfide dihydrochloride, dithiourea dihydrochloride, c2h6n4s2.2hcl, carbamimidoyldisulfanylmethanimidamide dihydrochloride, formamidine disulfide dihydrochloride disulfide dihydrochloride
PubChem CID 24802135
IUPAC Name [(azaniumylmethanimidoyl)disulfanyl](imino)methanaminium dichloride
SMILES [Cl-].[Cl-].[NH3+]C(=N)SSC([NH3+])=N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 223.13
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Formamidine Disulfide Dihydrochloride
RTECSNumber LQ4570000
TSCA Yes