missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formamide Solution, NMR Reference Standard, 90% in DMSO-d6 (99.9 atom% D), NMR tube size 5 mm 8 in., MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | 5 mm O.D. tube |
| CAS | 75-12-7 |
| Flash Point | 87.8°C |
| Linear Formula | CH3 NO |
| MDL Number | MFCD00007941 |
| Type | NMR Reference Standard |
| Quantity | 0.7 mL |
| Formula Weight | 45.04 |
| Grade | NMR Reference Standard |
| Concentration or Composition (by Analyte or Components) | 90% in DMSO- d6 (99.9 atom % D) |