missing translation for 'onlineSavingsMsg'
Learn More

Formamide Solution NMR Reference Standard, 45 in DMSO-d6 (99.9 atom D), NMR tube size 3 mm, MilliporeSigma™ Supelco™

Catalog No. 111002535
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002535 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002535 Supplier Merck Emd Millipore Supplier No. 6127071EA

Specifications

CAS 75-12-7
Concentration 45% in DMSO- d 6 (99.9 atom% D)
Linear Formula CH3 NO
MDL Number MFCD00007941
Type NMR Reference Standard
Quantity 1 Ea.
Formula Weight 45.04
Grade NMR Reference Standard