missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, stabilized with Methanol, ∼37 wt. in Water, MilliporeSigma™ Supelco™

Catalog No. 111016562
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016562 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016562 Supplier Merck Emd Millipore Supplier No. 47083U
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Ignition Point 572°F
CAS 50-00-0
Vapor Pressure 52 mmHg (37°C)
Vapor Density 1.03 (vs. air)
Density 1.09 g/mL (at 25°C)
Flash Point 56°C
Linear Formula HCHO
MDL Number MFCD00003274
Synonym Formalin
Type Food and Cosmetic Component Standard
Quantity 1 mL
Formula Weight 30.03
Grade Certified Reference Material
Concentration or Composition (by Analyte or Components) 37 wt. % aq. soln.
Show More Show Less