missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, min. 37% GR for Analysis, ACS, Reagent Ph Eur Grade, MilliporeSigma™
Supplier: Technidata 1040032500
Description
For analysisSpecifications
| Formaldehyde solution | |
| 50-00-0,67-56-1 | |
| min. 37% | |
| Plastic bottle | |
| MFCD00003274 | |
| UN2209 | |
| WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
| formaldehyde | |
| 712 | |
| ACS/Reagent Ph. Eur. |
| Stabilized with about 10% methanol | |
| ca.300°C | |
| Stinging | |
| CH2O | |
| 2.5 L | |
| formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
| C=O | |
| 30.03 | |
| CHEBI:16842 | |
| Liquid |
Chemical Identifiers
| 50-00-0 | |
| 30.03 | |
| WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
| 712 | |
| formaldehyde |
| CH2O | |
| MFCD00003274 | |
| formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
| CHEBI:16842 | |
| C=O |
Safety and Handling
H227: Combustible liquid.
H301 + H311 + H331: Toxic if swallowed
DOTInformation : DOT Class: 8, Packaging Group: III