missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, min. 37% Free From Acid, MilliporeSigma™

Catalog No. p-4939152
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Plastic bottle
Plastic container
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1039991000 1 L Plastic bottle
M1039992500 2.5 L Plastic bottle
M1039999025 25 L Plastic container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1039991000 Supplier Technidata Supplier No. 1.03999.1000
Only null left
For histology

Chemical Identifiers

CAS 50-00-0 , 67-56-1
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Ignition Point ca.300°C
Odor Stinging
Assay Percent Range min. 37%
Packaging Plastic bottle
Quantity 1 L
UN Number UN2209
Vapor Pressure 1.3 hPa
Physical Form Liquid
Chemical Name or Material Formaldehyde solution, Stabilized with about 10% methanol and calcium carbonate
Hazard Category H301 + H311 + H331: Toxic if swallowed
DOTInformation DOT Class: 8, Packaging Group: III