missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP, 35 wt. %, Solstice
Description
- Solstice has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | <-15°C |
| Color | Colorless |
| Density | 1.09g/mL at 25°C |
| pH | 3.5 to 4.5 (at 20°C) |
| Boiling Point | 93°C to 96°C |
| Flash Point | 64°C (147.2°F) |
| Ignition Residue | ≤0.01% (as SO4) |
| Packaging | Poly Bottle |
| Linear Formula | HCHO |
| Quantity | 2.5 L |
| Show More |
Safety and Handling
| Hazard Statement | P201-P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P308 + P311 |
| Precautionary Statement | H301 + H311 + H331-H314-H317-H335-H341-H350-H370 |
| DOTInformation | 8/PGIII |