missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, Contains 10-15% Methanol, Solstice

Catalog No. 60047995
Change view
Click to view available options
Quantity:
25 mL
500 mL
4 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047995 500 mL Poly Bottle
60047993 25 mL Poly Bottle
60047994 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60047995 Supplier Solstice Supplier No. F1635500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains 10-15% methanol as stabilizer, 37 wt. % in H2O

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point <-15°C
Color Colorless
Density 1.09g/mL at 25°C
pH 3.5 to 4.5 (at 20°C)
Boiling Point 93°C to 96°C (1013 hPa)
Flash Point 64°C (147.2°F)
Odor Stinging
Packaging Poly Bottle
Linear Formula HCHO
Quantity 500 mL
UN Number UN2209
Beilstein 1209228
Formula Weight 30.03g/mol
Chemical Name or Material Formaldehyde solution, 37 wt. % in H2O
Show More Show Less
Hazard Statement P201-P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P308 + P311
DOTInformation 8/PGIII