Learn More
New Pig Formaldehyde Neutralizing Absorbent Mat Pad
Minimize dangerous vapors and make formaldehyde spills safer to clean up!
Supplier:  New Pig MAT1400
Catalog No. 18-389-129
Only null left
Explore available promotions
Description
Description
  • Use pads as a surface liner, underneath containers or during transportation to neutralize formaldehyde vapors during a spill
  • Specially treated polypropylene quickly absorbs formaldehyde, formalin, formaldehyde-based fixatives, glutaraldehyde and other aldehyde solutions
  • Quickly neutralizes formaldehyde vapors to help protect workers
  • Durable top surface resists rips, tears and abrasion
  • Blue color helps distinguish mat from standard absorbents; provides contrast against tissue-based specimens in the medical field
  • High Btu value produces low ash when burned; ideal for fuels blending and incineration
Specifications
Specifications
Polypropylene
Product Suggestions
Product Suggestions