missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde dimethyl acetal, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020399
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020399 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020399 Supplier Merck Emd Millipore Supplier No. 887891MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 109-87-5
Density 0.860 g/mL (at 20°C)
Flash Point -18°C
Physical Form Neat
Linear Formula CH2(OCH3)2
MDL Number MFCD00008495
Boiling Point 41°C to 43°C
Synonym Dimethoxymethane; Methylal
Type Analytical Standard
Quantity 1 mL
Formula Weight 76.09
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less