missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, USP, Spectrum™ Chemical

Catalog No. p-7157477
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602835 125 mL Poly Bottle
18602834 500 mL Amber Glass Bottle
18602831 1 L Poly Bottle
18602832 4 L Poly Bottle
18602833 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602835 Supplier Spectrum Chemical Mfg Cor Supplier No. FO110125ML
Only null left

FO110, 50-00-0, HCHO

Spectrum™ Chemical Formaldehyde, 37 Percent, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 125 mL
UN Number UN1198
Percent Purity 34.5%
Grade USP