missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde 37%, StatLab™

37% Formaldehyde formula used to prepare formalin solutions.

$354.20 - $354.20

Specifications

CAS 50-00-0
Concentration or Composition (by Analyte or Components) 37% Formaldehyde, 63% Methyl Alcohol
View More Specs

Alert:

Harmful if swallowed, if in contact with skin, or if inhaled.


Products 1
Catalog Number Mfr. No. Packaging Price Quantity  
Catalog Number Mfr. No. Packaging Price Quantity  
STL285305
View Documents
Statlab
285305*
Cube
Each for $354.20
Only null left
 
Description

Description

  • Contains methanol as a stabilizer
Specifications

Specifications

50-00-0
37% Formaldehyde, 63% Methyl Alcohol
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'