missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 36.5 to 38%, Reagent grade, For ACS analysis, MP Biomedicals™

Catalog No. ICN19404780
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19404780 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19404780 Supplier MP Biomedicals Inc Supplier No. 0219404780

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Color Clear colorless
Density 1.04g/cm3 at 20°C
Quantity 100 mL
Formula Weight 30.03
Percent Purity 36.5 to 38%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Formaldehyde, For ACS analysis