Learn More
StatLab FormaGuard Neutralizing Pad
Provides an effective way for lab personnel to guard against formaldehyde vapors and exposure by allowing formalin penetration into a highly absorbent core.
Supplier:  StatLab FGP0811*
 View more versions of this product
Catalog No. STLFGP0811
Only null left
Description
Description
The FormaGuard™ Pad is light blue, rigid, polycoated, and roughly 1/8 in. in thickness
Specifications
Specifications
Pre-treated Formalin Neutralizer
Formalin Neutralizer
Pad
8 x 11 in.
Product Suggestions
Product Suggestions