missing translation for 'onlineSavingsMsg'
Learn More

Forchlorfenuron, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 456290010

 View more versions of this product

Catalog No. AC456290010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Forchlorfenuron
C12H10ClN3O
Solubility in water: 0.039g/L. Other solubilities: soluble in methanol (119g/L), ethanol (149g/L),, acetone (127g/L);, slightly soluble in chloroform (2.7g/L)
ClC1=CC(NC(=O)NC2=CC=CC=C2)=CC=N1
247.68
CHEBI:81861
≥97.5% (HPLC)
68157-60-8
MFCD00059898
GPXLRLUVLMHHIK-UHFFFAOYSA-N
1-(2-chloropyridin-4-yl)-3-phenylurea
93379
247.68

Chemical Identifiers

68157-60-8
247.68
GPXLRLUVLMHHIK-UHFFFAOYSA-N
CHEBI:81861
ClC1=CC(NC(=O)NC2=CC=CC=C2)=CC=N1
C12H10ClN3O
MFCD00059898
93379
1-(2-chloropyridin-4-yl)-3-phenylurea
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Suspected of causing cancer. Toxic to aquatic life with long lasting effects.

GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only