Learn More
Greenwich Safety For Responders QWIK-COVER™ Pre- and Post-Decon Dressing Kits
Maximize decontamination shower throughput by facilitating undress outside of the decon system
Supplier:  Greenwich Safety DCN001Y
Encompass
 View more versions of this product
Catalog No. 19161568
Only null left
Description
Description
Opaque decon garments fit over clothing, allowing ambulatory patients to disrobe in privacy. Perforations in these LLDPE plastic garments allow size adjustments in the overall length, arms, and head; as well as ensuring easy tear-away removal or conversion into a stretcher drape. Modesty flaps above the arm holes preserve privacy of the upper body. Color coded for easy identification of victim status. Packaged in easy-to-access zip-lock bags with pictographs for quick instructions.
Pre-Decon
  • One pre-decon garment One large SECUR-ID™ garment bag One small SECUR-ID valuables bag
Post-Decon
  • One post-decon garment 39 1/2 x 20 1/2 in. single-use Kimberly-Clark™ Dry-Up™ towel (amount varies for youth or large adult kits) Pair of spun bond polyethylene booties Molded nylon comb
Saddlebag Twin-Pack
  • One pre-decon kit One post-decon kit Connected in an easy-to-carry saddlebag design
Specifications
Chemical Identifiers
Dressing Kit
Specifications
Dressing Kit
Youth
Product Suggestions
Product Suggestions