Learn More
Greenwich Safety For Responders QWIK-COVER™ Pre- and Post-Decon Dressing Kits
Catalog No. p-2622834
Encompass
Change view
Click to view available options
Size:
Adult
Youth
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19161570 Adult
19161568 Youth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19161570 Supplier Greenwich Safety Supplier No. DCN001LA
Only null left
Maximize decontamination shower throughput by facilitating undress outside of the decon system
Opaque decon garments fit over clothing, allowing ambulatory patients to disrobe in privacy. Perforations in these LLDPE plastic garments allow size adjustments in the overall length, arms, and head; as well as ensuring easy tear-away removal or conversion into a stretcher drape. Modesty flaps above the arm holes preserve privacy of the upper body. Color coded for easy identification of victim status. Packaged in easy-to-access zip-lock bags with pictographs for quick instructions.
Pre-Decon
  • One pre-decon garment One large SECUR-ID™ garment bag One small SECUR-ID valuables bag
Post-Decon
  • One post-decon garment 39 1/2 x 20 1/2 in. single-use Kimberly-Clark™ Dry-Up™ towel (amount varies for youth or large adult kits) Pair of spun bond polyethylene booties Molded nylon comb
Saddlebag Twin-Pack
  • One pre-decon kit One post-decon kit Connected in an easy-to-carry saddlebag design
Specifications
Product Type Dressing Kit
Size Adult