missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid, calcium salt pentahydrate, 95.0-105.0%

Catalog No. AC230312500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC230312500 250 mg Glass bottle
AC230310010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC230312500 Supplier Thermo Scientific Chemicals Supplier No. 230312500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6035-45-6 | C20H21CaN7O7·5H2O | 601.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6035-45-6
Molecular Formula C20H21CaN7O7·5H2O
Molecular Weight (g/mol) 601.58
MDL Number MFCD00149465
InChI Key NPPBLUASYYNAIG-UHFFFAOYSA-L
Synonym folinic acid calcium pentahydrate
PubChem CID 131674093
IUPAC Name calcium;4-[[4-[(2-amino-5-formyl-4-oxido-7,8-dihydro-6H-pteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate;pentahydrate
SMILES C1C(N(C2=C(N1)N=C(N=C2[O-])N)C=O)CNC3=CC=C(C=C3)C(=O)NC(CCC(=O)[O-])C(=O)O.O.O.O.O.O.[Ca+2]

Specifications

Melting Point 240.0°C to 250.0°C
Color White-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 4249
Specific Rotation + 14.30
Solubility Information Solubility in water: freely soluble in water. Other solubilities: practically insoluble in alcohol
Formula Weight 601.58
Percent Purity 95 to 105%
Physical Form Powder
Chemical Name or Material Folinic acid, calcium salt pentahydrate, 95.0-105.0%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement AIR SENSITIVE
EINECSNumber 216-082-8
RTECSNumber MA0600500

RUO – Research Use Only