missing translation for 'onlineSavingsMsg'
Learn More

Medisca Folic Acid, USP (Dietary Supplement Grade)

Catalog No. 14186448
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186448 10 g
14186122 500 g
14185423 25 g
14185280 100 g
14185444 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14186448 Supplier Medisca Supplier No. 022401
Get 1 Extra Chemical When You Buy 4
View all promotions

missing translation for 'validMainframeMsg'

Folic Acid, USP (Dietary Supplement Grade) is an odorless, crystalline powder that can vary between yellow, yellow-brownish, or yellowish orange in color.

Folic Acid, USP is an odorless, crystalline powder that can vary between yellow, yellow-brownish, or yellowish orange in color.

Dietary Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered dietary supplement manufacturer or manufacturer with other certifications and registrations, based on applicable laws and regulations. Qualified manufacturers follow their applicable standards of practice, which are less stringent than those for FDA-registered drug manufacturers. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Folate, Vitamin B9, Pteroylglutamic acid

Specifications

Chemical Name or Material Folic Acid, USP (Dietary Supplement Grade)
Packaging Jar, 50mL, White, HDPE, Round, 38-400, TE Pano Neck
CAS 59-30-3
Type Drug Standard
Molecular Weight (g/mol) 441.4
Quantity 10 g
Percent Purity 97 to 102 %
Grade USP