missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid, Powder, USP, 97-102%, Spectrum™ Chemical

FO105, 59-30-3, C19H19N7O6

Supplier:  Spectrum Chemical Mfg Cor FO1051KG

 View more versions of this product

Catalog No. 18602805


Only null left

Description

Description

Spectrum™ Chemical Folic Acid, Powder, USP is a water soluble B vitamin used to treat folic acid deficiency associated with ulcerative colitis, alcoholism, kidney dialysis, and liver disease and certain types of anemia. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

59-30-3
0.3%
Amber Glass Bottle
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
1 kg
97 to 102%
100%
C19H19N7O6
OVBPIULPVIDEAO-LBPRGKRZSA-N
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
441.40
USP

Chemical Identifiers

59-30-3
441.40
(2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
C19H19N7O6
OVBPIULPVIDEAO-LBPRGKRZSA-N
NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions