missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid, Powder, USP, 97-102%, Spectrum™ Chemical

Catalog No. p-7157476
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602802 10 g Amber Glass Bottle
18602803 25 g Amber Glass Bottle
18602804 100 g Amber Glass Bottle
18602805 1 kg Amber Glass Bottle
18602806 5 kg Poly Pail
18602807 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18602802 Supplier Spectrum Chemical Mfg Cor Supplier No. FO10510GM

FO105, 59-30-3, C19H19N7O6

Spectrum™ Chemical Folic Acid, Powder, USP is a water soluble B vitamin used to treat folic acid deficiency associated with ulcerative colitis, alcoholism, kidney dialysis, and liver disease and certain types of anemia. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.40
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
IUPAC Name (2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
SMILES NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Packaging Amber Glass Bottle
Quantity 10 g
Percent Purity 97 to 102%
Grade USP