missing translation for 'onlineSavingsMsg'
Learn More

Folic acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014786
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014786 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014786 Supplier Merck Emd Millipore Supplier No. PHR10351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 59-30-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C19H19N7O6
MDL Number MFCD00079305
Synonym PteGlu; Pteroyl-L-glutamic acid; Vitamin M
Type Certified Reference Material
Quantity 1 g
Formula Weight 441.40
Grade Certified Reference Material