missing translation for 'onlineSavingsMsg'
Learn More

Folic acid dihydrate, 97%

Catalog No. AAA1430022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1430022 100 g
AAA1430014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1430022 Supplier Thermo Scientific Chemicals Supplier No. A1430022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75708-92-8 | C19H23N7O8 | 477.434 g/mol

Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid dehydrate is used to form non-covalent complexes with polyethylenimine. It is used for the delivery of genes, which finds application in gene therapy. It is a B-complex vitamin, which is necessary for the body to produce red blood cells.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 75708-92-8
Molecular Formula C19H23N7O8
Molecular Weight (g/mol) 477.434
MDL Number MFCD00079305
InChI Key ODYNNYOEHBJUQP-LTCKWSDVSA-N
Synonym folic acid dihydrate, unii-0s1t5c0jzl, 0s1t5c0jzl, pubchem18447, l-glutamic acid, n-4-2-amino-1,4-dihydro-4-oxo-6-pteridinyl methyl amino benzoyl-, dihydrate, l-glutamic acid, n-4-2-amino-3,4-dihydro-4-oxo-6-pteridinyl methyl amino benzoyl-, hydrate 1:2, folicacid,vitaminmorvitaminbc,vitaminm,pteglu, 2s-2-4-2-amino-4-hydroxypteridin-6-yl methyl amino phenyl formamido pentanedioic acid dihydrate, s-2-4-2-amino-4-oxo-3,4-dihydropteridin-6-yl methyl amino benzamido pentanedioic acid dihydrate
PubChem CID 16211651
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid;dihydrate
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N.O.O

Specifications

Melting Point ∼250°C (decomposition)
Quantity 100 g
Beilstein 100781
Merck Index 14,4221
Solubility Information Miscible with water.
Optical Rotation +18° (c=0.5 in 0.1N NaOH)
Formula Weight 477.44 (441.40 Anhydrous)
Percent Purity 97%
Chemical Name or Material Folic acid dihydrate
EINECSNumber 200-419-0
RTECSNumber LP5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only