missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Fold-ups Signs - Multiple Messages
Identify temporary hazardous conditions or display messages quickly and easily
$82.31
Specifications
| Material | Plastic |
|---|---|
| Quantity | 1 Ea. |
| For Use With (Application) | Identifying temporary hazardous conditions or displaying important messages, quickly and easily |
| Height (English) | 20 in. |
| Width (English) | 12 in. |
| Catalog Number | Mfr. No. | Displayed Text | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Displayed Text | Price | Quantity & Availability | |||||
|
191202790
|
Accuform Signs
PFR225 |
Do Not Enter (with international graphic) |
Each for $82.31
|
|
|||||
|
191202792
|
Accuform Signs
PFR227 |
Do Not Enter (with hand graphic) |
Each for $82.31
|
|
|||||
Description
- Double-sided signs can be set-up and moved easily to exactly where they are needed
- Rugged, high-impact plastic construction with handle cut-out
- Fold flat for easy storage and transport
- All signs include same text and pictogram on both sides
- All signs measure 20H x 12 in. W (51 x 30.5cm)
Specifications
| Plastic | |
| Identifying temporary hazardous conditions or displaying important messages, quickly and easily | |
| 12 in. | |
| Fold-up Floor Safety Sign |
| 1 Ea. | |
| 20 in. | |
| 12 x 20 in. |