Learn More
Contec™ FoamZorb™
Catalog No. 296367888
Change view
Click to view available options
FoamZorb™
Catalog No.
296367888 FoamZorb™
1 options
Catalog No. 296367888 Supplier Contec™ Supplier No. FZ451/8
FoamZorb™ foam wipes for use in cleanroom, strong open cell structure, combine high absorbency with low levels of extractable residue and particles, easy to wipe with for spill pickup, solvent and solution application and ideal for lithographic screen cleaning
  • Hydrophilic polyurethane foam for spill pickups
  • High sorptive capacity designed for solvent or solution applications
  • Excellent padding for reduction of vibration under sensitive products
Specifications
Material Polyurethane
Type Foam Wipes
For Use With (Application) Spill pickup, solvent and solution applications, lithographic screen cleaning