Learn More
Foamtec UltraSOLV™ ScrubPADS™
Vacuum Chamber Cleaning ScrubPADS for light to heavy duty PM procedures
Supplier:  Foamtec HT452210
Encompass
 View more versions of this product
Catalog No. 19042110
Only null left
Description
Description
Ideal for wafer processing tools such as LAM 9600 and 2300 Etch chambers, Hitachi Etch chambers, TEL Etch Chambers, Varian Ion Implant Process Areas, Novellus Speed, AMAT TxZ, WxZ, DxZ, SinGEN, PolyGEN, Endura and Producer CVD, PVD tools and Roth and Rau SiNX CVD tools.
  • Wide grit selection allows for quick removal of process-induced residue from aluminum, stainless steel, ceramic, glass, quartz and anodized surfaces while minimizing tool wear
  • Fiber-free construction reduces particle levels in cleaned tools
  • Unique bonding technology minimizes the release of abrasive particles leading to cleaner PM procedures
  • Enables the elimination of H2O2 from PMs, which greatly reduces recovery times
Specifications
Chemical Identifiers
Quick removal of process induced residue from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces while minimizing tool wear
Specifications
Vacuum Chamber Cleaning Scrub Pad
3 x 4 in.
Wafer processing tools
Without slit
10.16 cm
0.078 in.
3 x 4 in.
Quick removal of process induced residue from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces while minimizing tool wear
220
4 in.
Silicone Carbide
Product Suggestions
Product Suggestions