Learn More
Foamtec UltraSOLV™ ScrubDISKs
$73.98 - $8,205.10
Specifications
| Description | Fiber-free construction |
|---|---|
| For Use With (Application) | Quick removal of process induced residue from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces while minimizing tool wear |
| Material | Diamond |
| Catalog Number | Mfr. No. | Grit | Includes | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Grit | Includes | Price | Quantity & Availability | ||||
191202192
|
Foamtec
HT4580DC3 1 |
800 | Pressure-sensitive adhesive or hook and loop fastener |
Pack for $101.47
Case of 100 Pack for $8,027.00
|
|
||||
19304810
|
Foamtec
HT4530PDC3 1 |
3000 | Hook and loop fastener on handle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19304809
|
Foamtec
HT4520PDC3 1 |
2000 | Hook and loop fastener |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
19304851
|
Foamtec
HT4513PDC31 |
1350 | Hook and loop fastener |
Each for $73.98
Case of 100 Each for $6,148.00
|
|
||||
Description
Wide grit selection allows for quick removal of process-induced residue from aluminum, stainless steel, ceramic, glass, quartz and anodized surfaces and minimizes tool wear.
- Patented scrubbing fiber free tool
- Fiber-free construction for reducing particle levels in cleaned vacuum chambers
- Unique bonding technology for minimizing release of abrasive particles
- Eliminates H2O2 from PMs
- Removes hardened process residue from Hi vacuum chambers
- For use in conjunction with ErgoSCRUB™ hand tool
Specifications
| Fiber-free construction | |
| Quick removal of process induced residue from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces while minimizing tool wear | |
| Diamond |