Learn More
Foamtec UltraSOLV™ ScrubDISKs
Catalog No. 19304810
Encompass
Change view
Click to view available options
Grit:
1350
2000
3000
800
Includes:
Hook and loop fastener
Hook and loop fastener on handle
Pressure-sensitive adhesive or hook and loop fastener
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grit Includes
19304810 3000 Hook and loop fastener on handle
191202192 800 Pressure-sensitive adhesive or hook and loop fastener
19304809 2000 Hook and loop fastener
19304851 1350 Hook and loop fastener
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19304810 Supplier Foamtec Supplier No. HT4530PDC3 1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Wide grit selection allows for quick removal of process-induced residue from aluminum, stainless steel, ceramic, glass, quartz and anodized surfaces and minimizes tool wear.
  • Patented scrubbing fiber free tool
  • Fiber-free construction for reducing particle levels in cleaned vacuum chambers
  • Unique bonding technology for minimizing release of abrasive particles
  • Eliminates H2O2 from PMs
  • Removes hardened process residue from Hi vacuum chambers
  • For use in conjunction with ErgoSCRUB™ hand tool
Specifications
Product Type Scrub Disk
Description Fiber-free construction
Diameter (English) 3.5 in.
Diameter (Metric) 8.89 cm
Disposable Single-use
For Use With (Application) Quick removal of process induced residue from Aluminum, Stainless Steel, Ceramic, Glass, Quartz and Anodized surfaces while minimizing tool wear
For Use With (Equipment) ErgoScrub hand tool
Grit 3000
Includes Hook and loop fastener on handle
Material Diamond