Learn More
Foamtec Stainless Steel Corrosion Remediation Kits
Enable technicians to quickly remediate corrosion using a polishing and cleaning process that is fully compatible with cleanroom operations
$14.33 - $2,952.00
Specifications
| For Use With (Application) | Cleaning corrosion on stainless steel surfaces |
|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
19304555
|
Foamtec
HT4500 SSC3 |
Large Surface Starter Kit |
Each for $1,789.59
|
|
|||||
19304812
|
Foamtec
HT4500 DM1 |
Small Surface Starter Scrubpad Refill Kit |
Case of 10 Each for $2,952.00
|
|
|||||
19304581
|
Foamtec
HT4500 SSMNI |
Mini Kit |
Each for $920.92
|
|
|||||
|
19305775
|
Foamtec
HS4854 1 |
Natural Sahara Sponge |
Each for $14.33
Case of 100 Each for $1,191.00
|
|
|||||
Description
Kits include polishing and cleaning products along with instructional videos and documentation that result in mirror smooth 304 and 316 SS surfaces that resist subsequent corrosion.
Kit Contents
19-304-555: Stainless Steel Corrosion Remediation Starter Kit for Large Surfaces
2 Handles, 10 Diamond ScrubDISKS (5 girts), 1 Silicon-Carbide ScrubDISK, 1PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 25 MiraWIPE Wipers, 2 UltraSOLV Sponge, and Flash Drive with instructional Videos
19-304-556: Stainless Steel Corrosion Remediation Starter Kit
1 PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 10 MiraWIPE Wipers, 1 UltraSOLV Sponge, and Flash Drive with instructional Videos
Specifications
| Cleaning corrosion on stainless steel surfaces |