Learn More
Foamtec Stainless Steel Corrosion Remediation Kits
Catalog No. 19304555
Encompass
Change view
Click to view available options
Product Type:
Large Surface Starter Kit
Mini Kit
Natural Sahara Sponge
Small Surface Starter Scrubpad Refill Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19304555 Large Surface Starter Kit
19304812 Small Surface Starter Scrubpad Refill Kit
19304581 Mini Kit
19305775 Natural Sahara Sponge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19304555 Supplier Foamtec Supplier No. HT4500 SSC3
Enable technicians to quickly remediate corrosion using a polishing and cleaning process that is fully compatible with cleanroom operations
Kits include polishing and cleaning products along with instructional videos and documentation that result in mirror smooth 304 and 316 SS surfaces that resist subsequent corrosion.
Kit Contents
19-304-555: Stainless Steel Corrosion Remediation Starter Kit for Large Surfaces
2 Handles, 10 Diamond ScrubDISKS (5 girts), 1 Silicon-Carbide ScrubDISK, 1PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 25 MiraWIPE Wipers, 2 UltraSOLV Sponge, and Flash Drive with instructional Videos
19-304-556: Stainless Steel Corrosion Remediation Starter Kit
1 PilloWIPE, 4 Diamond ScrubPADS (4 girts), 1 Silicon-Carbide ScrubPAD, ScrubWRIGHT Pen with 2 Belts, 10 MiraWIPE Wipers, 1 UltraSOLV Sponge, and Flash Drive with instructional Videos
Specifications
Product Type Large Surface Starter Kit
For Use With (Application) Cleaning corrosion on stainless steel surfaces
For Use With (Equipment) Sahara Corrosion Cleaning System
Includes 280 Diamond grit ScrubPAD, 1350 Diamond grit ScrubPAD, 1500 Silicon Carbide grit ScrubPAD, 2000 Diamond grit ScrubPAD, 280 Diamond grit ScrubDISK™, 360 Diamond grit ScrubDISK™, 800 Diamond grit ScrubDISK<