Learn More
Gibco™ FoamAway™ Irradiated AOF (animal origin-free) Antifoaming Agent
Description
FoamAway™ Irradiated AOF is a ready-to-use antifoaming agent designed to offer high performance and convenience in combating foam in your cell culture system. It is prediluted, presterilized, and packaged to connect directly to your bioreactor with no further processing required.
Specifications
Specifications
| Classification | Animal Origin-Free |
| Content And Storage | Store at room temperature. Shake well before use. Do not filter. |
| Culture Type | Suspension Cell Culture |
| For Use With (Application) | Cell Culture |
| Product Line | Gibco |
| Product Type | Antifoaming Agent |
| Phenol Red Indicator | No Phenol Red |
| Quantity | 0.5 L in a 1 L bag |
| Shipping Condition | Room Temperature |
| Volume (Metric) | 500 ml |
| Show More |
Frequently Asked Questions (FAQs)
The target shipping percentage of simethicone in FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent is 3%. This supports the defoaming assay noted on the COA for the shelf life of the product.
FoamAway Irradiated AOF (animal origin-free) Antifoaming Agent is formulated to a 4.0% simethicone concentration during manufacture, representing the upper end of the product's concentration range, to account for loss of simethicone during processing.
The final concentration of simethicone may be variable in the finished product, but is ≤4.0%. The final product must pass a defoaming activity assay as a quality release criterion, ensuring defoaming activity throughout the 8-month shelf life.
Thermo Fisher Scientific conducted extended stability testing showing that FoamAway Irradiated AOF Antifoaming Agent remains within specification for 12 months from the date of irradiation. At the 13-month time point, multiple lots met the defoaming activity specification (≤ 15 seconds). Thus, 12 months is the supported shelf life based on validated stability data./
The connectors are male and female luer locks.
We recommend shaking to thoroughly mix the product before each addition. Approximately 1:1,000 dilution may be the midrange of the working concentration, about 30 ppm. The range may be about a 1:20,000 to 1:600 depending on the process. For an extended culture, we recommend re-adding material during the run as it is incorporated with the foaming components; we recommend adding as little as possible to keep the downstream processes functioning.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.