Learn More
STERIS™ Foam 240 Detergent
Foam 240 Acid-Based Cleaner is a powerful detergent for pharmaceutical, biotech, and cosmetic industries. It generates clinging foam, reducing labor costs, and effectively cleans inorganic soils. With excellent rinsing properties, it’s easy to validate and reliable.
$820.69 - $3,540.72
Specifications
| Product Type | Acid Cleaner |
|---|---|
| Color-coded | No |
| Form | Clear Liquid |
| pH | 1.9 (Diluted) |
| Scent | Unscented |
Description
Foam 240 Acid-Based Process and Research Cleaner is specially formulated to deliver effective foam cleaning in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
HOW FOAM 240 WORKS
Foam 240 Acid Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms. When applied with foam spray equipment, it generates a clinging foam to increase contact time and reduce labor costs.
WHY FOAM 240?
- Productivity - Reduces cleaning labor by increasing detergent contact time with heavy clinging foam when used with a foam applicator.
- Efficacy - Effectively cleans scale and other inorganic soils with a blend of acid, surfactants and other performance-enhancing ingredients.
- Cost Savings - Reduces water consumption and cleaning time due to superior rinsing characteristics.
- Easy to Validate - Facilitates cleaning validation with a support documentation including non-specific analytical methods, and toxicity reports.
- Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
- pH - 1% solution 8.9, typical
- Solubility - Complete
- Foam - Moderate to high
- Rinsing - Excellent
Specifications
| Acid Cleaner | |
| Clear Liquid | |
| Unscented |
| No | |
| 1.9 (Diluted) | |
| Non-sterile |