missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ Foam 240 Detergent

Catalog No. 17010169
Change view
Click to view available options
Size:
5 gal.
30 gal.
Container Type:
Keg
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17010169 5 gal. Pail
17010170 30 gal. Keg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17010169 Supplier STERIS™ Supplier No. 1D2405
Only null left

Foam 240 Acid-Based Cleaner is a powerful detergent for pharmaceutical, biotech, and cosmetic industries. It generates clinging foam, reducing labor costs, and effectively cleans inorganic soils. With excellent rinsing properties, it’s easy to validate and reliable.

Foam 240 Acid-Based Process and Research Cleaner is specially formulated to deliver effective foam cleaning in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW FOAM 240 WORKS

Foam 240 Acid Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms. When applied with foam spray equipment, it generates a clinging foam to increase contact time and reduce labor costs.

WHY FOAM 240?

  • Productivity - Reduces cleaning labor by increasing detergent contact time with heavy clinging foam when used with a foam applicator.
  • Efficacy - Effectively cleans scale and other inorganic soils with a blend of acid, surfactants and other performance-enhancing ingredients.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing characteristics.
  • Easy to Validate - Facilitates cleaning validation with a support documentation including non-specific analytical methods, and toxicity reports.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Physical Properties
  • pH - 1% solution 8.9, typical
  • Solubility - Complete
  • Foam - Moderate to high
  • Rinsing - Excellent

Specifications

Product Type Acid Cleaner
Color-coded No
Form Clear Liquid
pH 1.9 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.