missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-D-Arg(Pbf)-OH 98.0+%, TCI America™

Catalog No. p-7129953
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08751G 1 g
F08755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08751G Supplier TCI America Supplier No. F08751G

Chemical Identifiers

CAS 187618-60-6
Molecular Formula C34H40N4O7S
Molecular Weight (g/mol) 648.78
MDL Number MFCD00237010
InChI Key HNICLNKVURBTKV-UHFFFAOYNA-N
Synonym fmoc-d-arg pbf-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-yl sulfonyl carbamimidamido pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, ambotzfaa1305, fmoc-darg pbf-oh, pubchem12397, na-fmoc-n?-pbf-d-arginine, fmoc-n-guanidino-pbf-d-arginine, nalpha-fmoc-nomega-pbf-d-arginine
PubChem CID 46737258
IUPAC Name 5-{[amino(2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonamido)methylidene]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
SMILES CC1=C2CC(C)(C)OC2=C(C)C(C)=C1S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 648.78
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)
TSCA No
Recommended Storage Refrigerator